missing translation for 'onlineSavingsMsg'
Learn More

meso-Tetraphenylporphine, 97%

Product Code. 10259653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259653 25 g Glass bottle 25g
10533731 1 g Glass bottle 1g
10783171 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10259653 Supplier Thermo Scientific Acros Supplier No. 171210250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 917-23-7 | C44H30N4 | 614.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-23-7
Molecular Formula C44H30N4
Molecular Weight (g/mol) 614.75
MDL Number MFCD00011680
InChI Key AQPPOLXYUQPDOD-UHFFFAOYSA-N
Synonym tetraphenylporphyrin, 5,10,15,20-tetraphenylporphyrin, meso-tetraphenylporphine, tetraphenylporphine, 5,10,15,20-tetraphenylporphine, meso-tetraphenylporphyrin, 21h,23h-porphine, 5,10,15,20-tetraphenyl, 5,10,15,20-tetraphenyl-21h,23h-porphine, meso-tetraphenylporphyrine, tetraphenyl porphine
PubChem CID 70186
IUPAC Name 5,10,15,20-tetraphenyl-21,22-dihydroporphyrin
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Blue to Purple
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alcohol
Formula Weight 614.75
Percent Purity 97%
Physical Form Glistening Crystalline Powder
Chemical Name or Material meso-Tetraphenylporphine
Show More Show Less
compliance-icons
Product Identifier
  • meso-Tetraphenylporphine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.