missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol, 99%

Product Code. 10348983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348983 500 g Plastic bottle 500g
10050790 25 g Plastic bottle 25g
10090820 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10348983 Supplier Thermo Scientific Acros Supplier No. 117825000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-32-6
Molecular Formula C4H10O4
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004710
InChI Key UNXHWFMMPAWVPI-UHFFFAOYNA-N
Synonym erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
PubChem CID 222285
ChEBI CHEBI:17113
SMILES OCC(O)C(O)CO

Specifications

Melting Point 117.0°C to 121.0°C
Color White
Boiling Point 329.0°C to 331.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]2CH2OH
Quantity 500 g
Beilstein 01,525
Merck Index 15,3733
Solubility Information (10% in water) Clear colorless
IUPAC Name butane-1,2,3,4-tetrol
Formula Weight 122.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material meso-Erythritol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.