missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dimercaptosuccinic Acid, TRC

Product Code. 30370962
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370962 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370962 Supplier Toronto Research Chemicals Supplier No. TRCD44674050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material meso-2,3-Dimercaptosuccinic Acid
CAS 304-55-2
Molecular Formula C4H6O4S2
Quantity 50 g
Synonym Chemet, DIM-SA, DMS, DMSA, Ro 1-7977, Succimer, meso-Dimercaptosuccinic Acid, meso-2,3-Dimercaptosuccinic Acid, (R*,S*)-2,3-Dimercaptobutanedioic Acid
InChI Formula InChI=1S/C4H6O4S2/c5-3(6)1(9)2(10)4(7)8/h1-2,9-10H,(H,5,6)(H,7,8)/t1-,2+
SMILES OC(=O)[C@H](S)[C@H](S)C(=O)O
Molecular Weight (g/mol) 182.22
Formula Weight 181.9708
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.