missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dimercaptosuccinic acid, 98%

Product Code. 10206860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206860 25 g Glass bottle 25g
10053180 5 g Glass bottle 5g
10553894 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10206860 Supplier Thermo Scientific Acros Supplier No. 226670250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304-55-2 | C4H6O4S2 | 182.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-55-2
Molecular Formula C4H6O4S2
Molecular Weight (g/mol) 182.21
MDL Number MFCD00064799
InChI Key ACTRVOBWPAIOHC-XIXRPRMCSA-N
Synonym succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid
PubChem CID 2724354
ChEBI CHEBI:63623
IUPAC Name (2S,3R)-2,3-bis(sulfanyl)butanedioic acid
SMILES C(C(C(=O)O)S)(C(=O)O)S

Specifications

Melting Point 196°C to 198°C
Color White
Loss on Drying 1% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH(SH)CH(SH)CO2H
Quantity 25 g
Merck Index 15,8995
Formula Weight 182.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material meso-2, 3-Dimercaptosuccinic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.