missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dimercaptosuccinic acid, 97%

Product Code. p-7023897
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11429026

Brand: Thermo Scientific Alfa Aesar A17909.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304-55-2 | C4H6O4S2 | 182.208 g/mol

It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.

Solubility
Soluble in water, and ethanol (25 mg/ml, results in colorless to light yellow, clear).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-55-2
Molecular Formula C4H6O4S2
Molecular Weight (g/mol) 182.208
MDL Number MFCD00064799
InChI Key ACTRVOBWPAIOHC-XIXRPRMCSA-N
Synonym succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid
PubChem CID 2724354
ChEBI CHEBI:63623
IUPAC Name (2S,3R)-2,3-bis(sulfanyl)butanedioic acid
SMILES C(C(C(=O)O)S)(C(=O)O)S

Specifications

Melting Point ∼196°C (decomposition)
Quantity 1 g
Beilstein 1725150
Sensitivity Air Sensitive
Merck Index 14,8865
Solubility Information Soluble in water,and ethanol (25mg/ml,results in colorless to light yellow,clear).
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material meso-2,3-Dimercaptosuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.