missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific™

Product Code. 11475817
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475817 2500 g 2500g
11455817 100 g 100g
11428830 250 g 250g
11465817 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11475817 Supplier Thermo Scientific Alfa Aesar Supplier No. B24382.0E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-36-6
Molecular Formula C4H2Br2O4
Molecular Weight (g/mol) 273.87
MDL Number MFCD00066439
InChI Key FJWGRXKOBIVTFA-XIXRPRMCSA-L
Synonym 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
PubChem CID 641611
IUPAC Name (2S,3R)-2,3-dibromobutanedioic acid
SMILES [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

Specifications

Melting Point ∼280°C
Quantity 2500 g
UN Number UN3261
Beilstein 1725173
Merck Index 14,3029
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.
Formula Weight 275.89
Percent Purity 98%
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.