missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Dibromo-1,2-diphenylethane, 97%

Product Code. p-7044166
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11361777

Brand: Thermo Scientific Alfa Aesar L04956.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13440-24-9 | C14H12Br2 | 340.058 g/mol

meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13440-24-9
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.058
MDL Number MFCD00000137
InChI Key GKESIQQTGWVOLH-LSLKUGRBSA-N
Synonym meso-dibromostilbene
PubChem CID 102063597
IUPAC Name [(1S)-1,2-dibromo-2-phenylethyl]benzene
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br

Specifications

Melting Point ∽241°C (decomposition)
Quantity 5 g
UN Number UN1759
Beilstein 2050820
Solubility Information Insoluble in water.
Formula Weight 340.07
Percent Purity 97%
Chemical Name or Material meso-1,2-Dibromo-1,2-diphenylethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.