missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Bis(p-tolyl)ethylenediamine, 98%, Thermo Scientific™

Product Code. 10795613 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795613 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795613 Supplier Thermo Scientific Acros Supplier No. 375271000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 50764-59-5
Melting Point 113°C to 117°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H20N2
Linear Formula (CH3C6H4CH(NH2)-)2
Quantity 100 mg
Synonym 1r,2s-1,2-bis 4-methylphenyl ethane-1,2-bis aminium, 1s,2r-2-azaniumyl-1,2-bis 4-methylphenyl ethyl azanium
InChI Key UQUZLWQGLCLOBD-IYBDPMFKSA-P
SMILES CC1=CC=C(C=C1)C(C(C2=CC=C(C=C2)C)[NH3+])[NH3+]
IUPAC Name [(1R,2S)-2-azaniumyl-1,2-bis(4-methylphenyl)ethyl]azanium
Molecular Weight (g/mol) 240.35
PubChem CID 7017343
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material meso-1, 2-Bis(p-tolyl)ethylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • meso-1,2-Bis(p-tolyl)ethylenediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.