missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Bis(4-bromophenyl)ethanediamine, 98+%, Thermo Scientific™

Product Code. 10250223 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 10250223

Brand: Thermo Scientific Acros 380461000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 117903-53-4
Color Green-Yellow to Pink
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C14H14Br2N2
MDL Number MFCD00989104
Quantity 100 mg
Synonym 1,2-bis 4-bromophenyl ethane-1,1-diamine, meso-1,2-bis 4-bromophenyl ethanediamine
InChI Key GANMXFWDVICUEK-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CC(C2=CC=C(C=C2)Br)(N)N)Br
IUPAC Name 1,2-bis(4-bromophenyl)ethane-1,1-diamine
Molecular Weight (g/mol) 370.09
PubChem CID 91872995
Formula Weight 370.09
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material meso-1, 2-Bis(4-bromophenyl)ethanediamine
Show More Show Less
compliance-icons
Product Identifier
  • meso-1,2-Bis(4-bromophenyl)ethanediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.