missing translation for 'onlineSavingsMsg'
Learn More

Mesitylenesulfonic acid sodium salt hemihydrate, 98%

Product Code. 11419517
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11419517

Brand: Thermo Scientific Alfa Aesar B22029.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6148-75-0 | C9H11NaO3S | 222.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6148-75-0
Molecular Formula C9H11NaO3S
Molecular Weight (g/mol) 222.23
MDL Number MFCD00067712
InChI Key AOJUNZYQOYSGHT-UHFFFAOYSA-M
Synonym sodium mesitylenesulfonate, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt, sodium 2,4,6-trimethylbenzenesulfonate, sodium 2,4,6-trimethylbenzenesulphonate, mesitylenesulfonic acid sodium salt hemihydrate, mesitylenesulfonic acid sodium salt, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1, 2,4,6-trimethylbenzenesulfonic acid sodium salt, mesitylene sodium sulfonate, acmc-1bbae
PubChem CID 23661859
IUPAC Name sodium;2,4,6-trimethylbenzenesulfonate
SMILES [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 5784990
Formula Weight 231.26 (222.25 Anhydrous)
Percent Purity 98%
Chemical Name or Material Mesitylenesulfonic acid sodium salt hemihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.