missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene, TRC

Product Code. 30475964
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475964 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475964 Supplier Toronto Research Chemicals Supplier No. TRCM258390500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3,5-Trimethylbenzene
CAS 108-67-8
Molecular Formula C9 H12
Quantity 500 mL
Synonym Benzene, 1,3,5-trimethyl-, Mesitylene (8CI), 1,3,5-Trimethylbenzene, 2,4,6-Trimethylbenzene, 3,5-Dimethyltoluene, NSC 9273, sym-Trimethylbenzene
InChI Formula InChI=1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3
SMILES Cc1cc(C)cc(C)c1
IUPAC Name 1,3,5-trimethylbenzene
Molecular Weight (g/mol) 120.19
Formula Weight 120.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.