missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals
20.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis
Solubility
Soluble in toluene (almost transparency).
Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis
Solubility
Soluble in toluene (almost transparency).
Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
Specifications
Melting Point | 53°C to 57°C |
Boiling Point | 150°C (20 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Irritating |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 1107601 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in toluene (almost transparency). |
Formula Weight | 218.7 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'