missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Oxide, TRC

Product Code. 30480787
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480787 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480787 Supplier Toronto Research Chemicals Supplier No. TRCM225635100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesityloxide
CAS 141-79-7
Molecular Formula C6 H10 O
Quantity 100 mL
Synonym 4-Methylpent-3-en-2-one, Cilastatin Sodium Imp. D (EP), 3-Penten-2-one, 4-methyl- (8CI, 9CI, ACI), 4-Methyl-3-penten-2-one (ACI), 2,2-Dimethylvinyl methyl ketone, 2-Methyl-2-penten-4-one, 2-Methyl-4-oxo-2-pentene, 4-Methyl-3-pentene-2-one, Isobutenyl methyl ketone, Isopropylideneacetone, Mesityl oxide, Methyl 2,2-dimethylvinyl ketone, Methyl 2-methyl-1-propenyl ketone, Methyl isobutenyl ketone, NSC 38717
InChI Formula InChI=1S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3
SMILES CC(=CC(=O)C)C
IUPAC Name 4-methylpent-3-en-2-one
Molecular Weight (g/mol) 98.14
Formula Weight 98.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.