missing translation for 'onlineSavingsMsg'
Learn More

Mesitaldehyde, 97%

Product Code. 11458830
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458830 2 g 2g
11499332 10 g 10g
11409342 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458830 Supplier Thermo Scientific Alfa Aesar Supplier No. A10104.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 487-68-3 | C10H12O | 148.21 g/mol

Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

Solubility
Soluble in Chloroform

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-68-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00003341
InChI Key HIKRJHFHGKZKRI-UHFFFAOYSA-N
Synonym mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
PubChem CID 10254
IUPAC Name 2,4,6-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C(C)=C1

Specifications

Melting Point 14°C
Density 1.005
Boiling Point 237°C
Flash Point 106°C (222°F)
Refractive Index 1.554
Quantity 2 g
Beilstein 1364114
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material Mesitaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.