missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MesenCult™ Proliferation Kit (Human)

Product Code. 17198261
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17198261 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17198261 Supplier STEMCELL Technologies Supplier No. 05411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for detection of CFU-F and expansion of human mesenchymal stem cells

  • MesenCult™ Proliferation Kit (Human) is a standardized, serum-containing medium for the culture of human mesenchymal stem cells (MSCs)
  • MesenCult™ Proliferation Kit (Human) is optimized for the expansion of human MSCs in vitro as well as for the detection and enumeration of colony-forming unit - fibroblasts (CFU-F)
  • MesenCult™ Proliferation Kit (Human) includes MesenCult™ MSC Basal Medium (Human; 450 mL) and MesenCult™ MSC Stimulatory Supplement (Human; 50 mL)

Specifications

Cell Type Mesenchymal Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Expansion
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.