missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MesenCult™ Adipogenic Differentiation Kit (Mouse)
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MesenCult™ Adipogenic Differentiation Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs), adipose tissue-derived MSCs (ADSCs), and mouse embryonic fibroblasts (MEFs) into cells of the adipogenic lineage
- NOTE: MesenCult™ Adipogenic Differentiation Medium must be supplemented with L-Glutamine (Catalog #07100)
Specifications
Specifications
| Cell Type | Mesenchymal Stem and Progenitor Cells |
| For Use With (Application) | Cell Differentiation |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction