missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MesenCult™ Adipogenic Differentiation Kit (Human)
Description
- MesenCult™ Adipogenic Differentiation Medium (Human) is specifically formulated for the in vitro differentiation of human mesenchymal stromal cells (also known as mesenchymal stem cells or MSCs) into adipogenic lineage cells
- This kit is suitable for the differentiation of MSCs derived from human bone marrow (BM), adipose tissue, umbilical cord, or pluripotent stem cells (PSCs) that have been previously culture-expanded in serum- and animal component-free medium (e.g. MesenCult™ ACF Plus Medium [Catalog #05445]), serum-containing medium (e.g. MesenCult™ Proliferation Kit [Catalog #05411]), or platelet lysate medium (e.g. MesenCult™-hPL Medium [Catalog #05439])
Specifications
Specifications
| Cell Type | Adipocytes, Mesenchymal Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?