missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MesenCult™-ACF Plus Culture Kit

Product Code. 17128271
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17128271 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17128271 Supplier STEMCELL Technologies Supplier No. 05448
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Animal component-free medium and attachment substrate for human mesenchymal stem cells

  • Reduce variability in your human mesenchymal stromal cell (MSC; also known as mesenchymal stem cell) cultures and improve experimental reproducibility by using this animal component-free (ACF) and extracellular vesicle (EV)-free medium
  • MesenCult-ACF Plus Medium is optimized so you can derive human MSCs from multiple sources, such as bone marrow or adipose tissue, without serum
  • Compared to serum-containing or EV-depleted serum-containing media, MSCs cultured with this kit expand more efficiently (rate and cumulative cell total) and without compromising function
  • The cultured cells exhibit characteristic MSC surface marker expression and retain robust expansion rate and trilineage differentiation capacities
  • This kit is part of a complete ACF workflow for deriving, expanding, and cryopreserving MSCs, as well as differentiating human pluripotent stem cells into mesenchymal progenitors that is optimized for efficient and consistent MSC cultures

Specifications

Cell Type Mesenchymal Cells, PSC-Derived, Mesenchymal Stem and Progenitor Cells
For Use With (Application) Cell Culture, Expansion, Maintenance
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.