missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MesenCult™-ACF Freezing Medium

Product Code. 16230521
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16230521 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16230521 Supplier STEMCELL Technologies Supplier No. 05490
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Animal component-free MSC cryopreservation medium

  • MesenCult™-ACF Freezing Medium is a defined, serum-free, and animal component-free medium for the cryopreservation of mesenchymal stem cells (MSCs)
  • This complete and ready-to-use medium is recommended for human MSCs previously cultured in MesenCult™-ACF Plus Medium (Catalog #05445) or MesenCult™ Medium (MesenCult™ Proliferation Kit; Catalog #05411)
  • Frozen human MSCs should be stored at -135°C (liquid nitrogen) or colder

Specifications

Cell Type Mesenchymal Stem and Progenitor Cells
For Use With (Application) Cryopreservation
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.