missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MesenCult™-ACF Chondrogenic Differentiation Kit

Product Code. 17138271
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138271 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17138271 Supplier STEMCELL Technologies Supplier No. 05455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal component-free medium for the differentiation of MSCs into chondrocytes

  • MesenCult™-ACF Chondrogenic Differentiation Medium is animal component-free (ACF) and specifically formulated for the in vitro differentiation of human mesenchymal stromal cells (MSCs; also known as mesencyhymal stem cells) into chondrogenic lineage cells, including chondrocytes
  • This medium is suitable for the differentiation of human bone marrow (BM)-, adipose tissue (AT)- and synovium (S)-derived MSCs previously culture-expanded in serum-containing medium (e.g. MesenCult™ Proliferation Kit [Catalog #05411]) or animal component-free MesenCult™-ACF Plus Medium [Catalog #05445])
  • MesenCult™-ACF Chondrogenic Differentiation Medium induces robust chondrogenic differentiation of human MSCs with as few as 3 x 10^5 cells and as early as day 14

Specifications

Cell Type Chondrocytes, Mesenchymal Stem and Progenitor Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.