missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MESDC2 Antibody (502), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89719H
This item is not returnable.
View return policy
Description
MESDC2 Monoclonal specifically detects MESDC2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MESDC2 | |
| Monoclonal | |
| HRP | |
| BOCA, KIAA0081Renal carcinoma antigen NY-REN-61, MESDLDLR chaperone MESD, mesoderm development candidate 2Mesoderm development protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MESDC2 (NP_055969.1; Ala 34-Lys 230). | |
| 0.1 mL | |
| Signal Transduction | |
| 23184 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 502 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction