missing translation for 'onlineSavingsMsg'
Learn More

MESDC2 Antibody (502), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18352641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352641 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352641 Supplier Novus Biologicals Supplier No. NBP289719AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MESDC2 Monoclonal specifically detects MESDC2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MESDC2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 502
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias BOCA, KIAA0081Renal carcinoma antigen NY-REN-61, MESDLDLR chaperone MESD, mesoderm development candidate 2Mesoderm development protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human MESDC2 (NP_055969.1; Ala 34-Lys 230).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23184
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.