missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES-SDS running buffer (20X)

Product Code. 15444569
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 15444569

Brand: Thermo Scientific Chemicals J62138.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.

Solubility
Miscible with water.

Notes
This buffer contains: 1M MES, 1M Tris base,2% SDS and 20mM EDTA at pH 7.3. Store at room temperature. Keep away from light. Incompatible with strong oxidizing agent and acids.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Physical Form Liquid
Quantity 1 L
compliance-icons
Product Identifier
  • MES-SDS running buffer (20X)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Morpholineethanesulfonic acid, monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.