Learn More
Thermo Scientific Chemicals MES-SDS running buffer (20X)
Description
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.
Solubility
Miscible with water.
Notes
This buffer contains: 1M MES, 1M Tris base,2% SDS and 20mM EDTA at pH 7.3. Store at room temperature. Keep away from light. Incompatible with strong oxidizing agent and acids.
Specifications
Specifications
| Concentration | 20X |
| Physical Form | Liquid |
| Quantity | 500 mL |
Safety and Handling
- MES-SDS running buffer (20X)
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium lauryl sulfate, EDTA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.