missing translation for 'onlineSavingsMsg'
Learn More

MES monohydrate, free acid, ≥99%, MP Biomedicals™

Product Code. 11466581 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
This item is not returnable. View return policy

Product Code. 11466581

Brand: MP Biomedicals 0219483583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa ± 1). Alternatively, volumes of equimolar MES free acid and sodium or potassium MES can be mixed to attain the desired pH

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-[N-morpholino]ethanesulfonic acid
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range ≥99%
Formula Weight 213.26
Percent Purity 99.6%
Chemical Name or Material MES monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.