missing translation for 'onlineSavingsMsg'
Learn More

MES monohydrate free acid ≥ 98%, crystalline, Molecular biology grade, J.T. Baker™

Product Code. 15598204 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15598204 1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598204 Supplier JT Baker Supplier No. 401404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥ 98%, crystalline, Ultrapure Bioreagent for liquid chromatography

  • Buffer for liquid chromatography and other molecular biology applications.
  • Lot analysis on label.
  • For Laboratory, Research, or Manufacturing Use
  • pKa at 20°C = 6.15

Specifications

CAS 145224-94-8
Melting Point 300 deg;C
Quantity 1 kg
Assay Percent Range ≥ 98% (C6H13NO4S·H2O)
Loss on Drying 7 - 10% (at 130°C)
Molecular Formula C6H13NO4S
MDL Number MFCD00149409
Packaging White poly bottle, blue tamper evident cap
Synonym 2-(N-Morpholino)ethanesulphonic acid monohydrate
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES [O-]S(=O)(=O)CC[NH+]1CCOCC1
IUPAC Name 4-(2-sulfonatoethyl)morpholin-4-ium
Molecular Weight (g/mol) 195.23
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.