missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry

Product Code. 10297403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297403 2.5 kg Plastic bottle 2.5kg
10517241 100 g Plastic bottle 100g
10781804 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10297403 Supplier Thermo Scientific Chemicals Supplier No. 327760025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 145224-94-8 | C6H15NO5S | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic acid monohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Absorbance (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point >300.0°C
Color Orange-Yellow to White
pH 2.5 to 4.0 (1%, H2O, 25°C)
Physical Form Crystals, Flakes or Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Information Solubility in water: soluble.
Formula Weight 213.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material MES monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.