missing translation for 'onlineSavingsMsg'
Learn More

MES Monohydrate, ≥98.5% (Dry Basis), Ultrapure

Product Code. 15835178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15835178 100 g Plastic bottle 100g
15875318 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15835178 Supplier Thermo Scientific Supplier No. J18886.22
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point ∼308°C (decomposition)
Packaging Plastic bottle
Linear Formula HO3S(CH2)2(C4H8NO)·H2O
Quantity 100 g
Beilstein 141319
Formula Weight 213.25
Percent Purity ≥98.5%
Grade Ultrapure
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material MES Monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.