missing translation for 'onlineSavingsMsg'
Learn More

MES lysis buffer with Triton™ X-100, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62693.AK

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15435019

  • £44.30 / 250mL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

MES lysis buffer with Triton™ X-100
Colorless
Liquid
250 mL
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers