missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES hydrate, 99+%, for biochemistry

Product Code. 10052190 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10052190

Brand: Thermo Scientific Chemicals 172591000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 4-Morpholinoethanesulfonic acid hydrate, MES
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1

Specifications

Absorbance (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.) (1 cm cell)
Melting Point 243.0°C
Color White
pH 2.5 to 4.0 (1%, H2O, 25°C)
Physical Form Needle-like Crystals or Powder
Quantity 100 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 9.5% max. (105°C, 3 hrs)
Packaging Plastic bottle
Merck Index 14, 5902
Solubility Information Solubility in water: soluble.
Formula Weight 195.23
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material MES hydrate, 0.99
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.