missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MES 50mM, NaCl 200mM

Product Code. 30484973
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
missing translation for 'unitSize'
100 L
20 L
200 L
5 L
50 L
This item is not returnable. View return policy

Product Code. 30484973

missing translation for 'mfr': Gibco™ A59170BA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Features

  • Available in a range of pack sizes
  • Established shelf life; set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
TRUSTED_SUSTAINABILITY

Specifications

Form Clear, colorless solution
Product Type Sodium Chloride
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Formulation MES/NaCl Buffer
Manufacturing Quality CGMP
Quantity 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.