missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 7.5

Product Code. 15495059
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495059 500 mL 500mL
15485059 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495059 Supplier Thermo Scientific Chemicals Supplier No. J62752.AP
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

Solubility
Soluble in water (0.5 M at 20°C).

Notes
Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2 mm filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Concentration 1.0 M
pH 7.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water (0.5 M at 20°C).
Chemical Name or Material MES buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.