missing translation for 'onlineSavingsMsg'
Learn More

MES, 0.5M buffer soln., pH 7.0, Thermo Scientific™

Catalog Number 15415359
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
This item is not returnable. View return policy
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.5 M
pH 7.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material MES buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'