missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MERTK (cMER), GST Tag Recombinant Protein
Human MERTK (cMER), GST Tag Recombinant Protein
Brand: Invitrogen™ PV3627
This item is not returnable.
View return policy
Description
MERTK (cMER) is a tyrosine kinase proto-oncogene and is involved in the retinal pigment epithelium (RPE) phagocytosis pathway, which is implicated in human retinal disease.Specifications
NP_006334 | |
10461 | |
10 μg | |
AXL Kinase Family | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
MERTK | |
Baculovirus | |
Recombinant, Fragment, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
63.4 kDa | |
MERTK (cMER), MER | |
aa 578-872 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
c-mer proto-oncogene tyrosine kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |