missing translation for 'onlineSavingsMsg'
Learn More

Mertansine, MedChemExpress

Product Code. 16460326
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460326

Brand: Medchem Express HY19792

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mertansine (DM1) is a microtubulin inhibitor and is an antibody-conjugatable maytansinoid that is developed to overcome systemic toxicity associated with maytansine and to enhance tumor-specific delivery. Mertansine can be attached to a monoclonal antibody with a linker to create an antibody-drug conjugate (ADC).

Mertansine (DM1) is a microtubulin inhibitor and is an antibody-conjugatable maytansinoid that is developed to overcome systemic toxicity associated with maytansine and to enhance tumor-specific delivery. Mertansine can be attached to a monoclonal antibody with a linker to create an antibody-drug conjugate (ADC).

Specifications

Chemical Name or Material Mertansine
CAS 139504-50-0
Color White
Purity Grade Notes Research
Molecular Formula C35H48ClN3O10S
Quantity 2 mg
Synonym DM1 Maytansinoid DM1
Solubility Information DMSO : 62.5 mg/mL (84.66 mM; Need ultrasonic)
SMILES C[C@]1([C@@](CC(N(C(C=C2C=C3OC)=C3Cl)C)=O)([H])OC([C@H](C)N(C)C(CCS)=O)=O)[C@H]([C@@H]([C@](OC4=O)([H])C[C@]([C@](/C=C/C=C(C)/C2)([H])OC)(N4)O)C)O1
Molecular Weight (g/mol) 738.29
Formula Weight 738.29
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.