missing translation for 'onlineSavingsMsg'
Learn More

MERS-CoV Spike Protein Antibody, DyLight 755, Novus Biologicals™

Product Code. 18327686 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327686 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327686 Supplier Novus Biologicals Supplier No. NBP305814IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MERS-CoV/NCov Spike Protein Polyclonal specifically detects MERS-CoV/NCov Spike Protein in MERS-CoV samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MERS-CoV/NCov Spike Protein
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coronavirus s1, coronavirus s2, coronavirus spike, cov spike, E2, Human coronavirus spike glycoprotein, MERS, MERS Peplomer protein, MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV E2, MERS-CoV Peplomer protein, MERSCoV S glycoprotein, MERS-CoV S glycoprotein, MERSCoV Spike glycoprotein, MERS-CoV Spike glycoprotein, MERSCOV Spike Protein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, ncov RBD, ncov s1, ncov s2, ncov spike, Peplomer proteinMERS-CoV Spike glycoprotein, RBD, S, S1, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, a synthetic peptide corresponding to the center region of the S1 subunit of MERS-CoV/NCov Spike Protein (S protein).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14254594
Target Species MERS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.