missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike Protein Antibody (3873) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05714
This item is not returnable.
View return policy
Description
MERS-CoV Spike Protein Monoclonal antibody specifically detects MERS-CoV Spike Protein in MERS-CoV samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| MERS-CoV Spike Protein | |
| Monoclonal | |
| Unconjugated | |
| MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
| Synthetic peptide | |
| 50 μg | |
| Infections (Virus Bacteria and Parasites) | |
| 14254594 | |
| Store at -20°C. | |
| IgG2a |
| ELISA, Immunofluorescence | |
| 3873 | |
| 0.01M PBS, pH7.2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| MERS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction