missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS Coronavirus Spike Protein S1 Rabbit anti-Virus, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA581785
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for MERS-CoV (NCoV/Novel coronavirus) Spike Protein S1.
Specifications
| MERS Coronavirus Spike Protein S1 | |
| Polyclonal | |
| PBS with no preservative | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| MERS Coronavirus S protein; Spike glycoprotein | |
| Recombit MERS-CoV (NCoV/Novel coronavirus) Spike Protein S1 Protein | |
| 100 μL | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction