missing translation for 'onlineSavingsMsg'
Learn More

Meropenem trihydrate, MedChemExpress

Product Code. 16481957
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481957 50 mg 50mg
16456276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481957 Supplier Medchem Express Supplier No. HY13678A
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Meropenem trihydrate (SM 7338 trihydrate) is a carbapenem antibiotic with broad-spectrum antibacterial activity. Meropenem trihydrate has activity against susceptible and resistant N. gonorrhoeae (MIC value of 0.02-0.06 mg/mL), H. influenzae (MIC value of 0.03-0.12 mg/mL), and H. ducreyi (MIC value of 0.015-0.12 mg/mL).

Meropenem trihydrate (SM 7338 trihydrate) is a carbapenem antibiotic with broad-spectrum antibacterial activity. Meropenem trihydrate has activity against susceptible and resistant N. gonorrhoeae (MIC value of 0.02-0.06 mg/mL), H. influenzae (MIC value of 0.03-0.12 mg/mL), and H. ducreyi (MIC value of 0.015-0.12 mg/mL).

Specifications

Chemical Name or Material Meropenem trihydrate
Physical Form Powder
CAS 119478-56-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H31N3O8S
For Use With (Application) COVID-19-immunoregulation
Synonym SM 7338 trihydrate
Solubility Information DMSO : 100 mg/mL (228.57 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (28.57 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(S[C@@H]3CN[C@H](C(N(C)C)=O)C3)[C@H](C)[C@]2([H])[C@@H]([C@H](O)C)C1=O)O.O.O.O
Molecular Weight (g/mol) 437.51
Formula Weight 437.51
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.