missing translation for 'onlineSavingsMsg'
Learn More

Mericitabine, MedChemExpress

Product Code. 16460117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460117 50 mg 50mg
16426467 5 mg 5mg
16403738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460117 Supplier Medchem Express Supplier No. HY10240
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mericitabine (RG 7128; R-7128) is a nucleoside inhibitor of the HCV NS5B polymerase that acts as an RNA chain terminator and prevents elongation of RNA transcripts during replication.

Mericitabine (RG 7128

Specifications

Chemical Name or Material Mericitabine
CAS 940908-79-2
Color White
Purity Grade Notes Research
Molecular Formula C18H26FN3O6
Quantity 50 mg
Synonym RG 7128 R-7128 PSI 6130 diisobutyrate
Solubility Information DMSO : 100 mg/mL (250.37 mM; Need ultrasonic)
SMILES O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2OC(C(C)C)=O)F)O[C@@H]2COC(C(C)C)=O)N
Molecular Weight (g/mol) 399.41
Formula Weight 399.41
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.