missing translation for 'onlineSavingsMsg'
Learn More

Merestinib, MedChemExpress

Product Code. p-200098090
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486786 100 mg 100mg
16482677 50 mg 50mg
16459157 5 mg 5mg
16456588 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486786 Supplier Medchem Express Supplier No. HY15514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Merestinib (LY2801653) is a potent, orally bioavailable c-Met inhibitor (Ki=2 nM) with anti-tumor activities. Merestinib (LY2801653) also has potent activity against MST1R (IC50=11 nM), FLT3 (IC50=7 nM), AXL (IC50=2 nM), MERTK (IC50=10 nM), TEK (IC50=63 nM), ROS1, DDR1/2 (IC50=0.1/7 nM) and MKNK1/2 (IC50=7 nM).

Merestinib (LY2801653) is a potent, orally bioavailable c-Met inhibitor (Ki=2 nM) with anti-tumor activities. Merestinib (LY2801653) also has potent activity against MST1R (IC50=11 nM), FLT3 (IC50=7 nM), AXL (IC50=2 nM), MERTK (IC50=10 nM), TEK (IC50=63 nM), ROS1, DDR1/2 (IC50=0.1/7 nM) and MKNK1/2 (IC50=7 nM).

Specifications

Chemical Name or Material Merestinib
CAS 1206799-15-6
Color White
Purity Grade Notes Research
Molecular Formula C30H22F2N6O3
Quantity 100 mg
Synonym LY2801653
Solubility Information DMSO : ≥ 32 mg/mL (57.92 mM)
SMILES O=C(C1=CC=C(C)N(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=CC5=C(N(C)N=C5)C=C4C6=CNN=C6)C(F)=C3
Molecular Weight (g/mol) 552.53
Formula Weight 552.53
Percent Purity 98.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.