missing translation for 'onlineSavingsMsg'
Learn More

Merestinib dihydrochloride, MedChemExpress

Product Code. 16496786
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496786

Brand: Medchem Express HY15514A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Merestinib dihydrochloride (LY2801653 dihydrochloride) is a potent, orally bioavailable c-Met inhibitor (Ki=2 nM) with anti-tumor activities. Merestinib dihydrochloride also has potent activity against MST1R (IC50=11 nM), FLT3 (IC50=7 nM), AXL (IC50=2 nM), MERTK (IC50=10 nM), TEK (IC50=63 nM), ROS1, DDR1/2 (IC50=0.1/7 nM) and MKNK1/2 (IC50=7 nM).

Merestinib dihydrochloride (LY2801653 dihydrochloride) is a potent, orally bioavailable c-Met inhibitor (Ki=2 nM) with anti-tumor activities. Merestinib dihydrochloride also has potent activity against MST1R (IC50=11 nM), FLT3 (IC50=7 nM), AXL (IC50=2 nM), MERTK (IC50=10 nM), TEK (IC50=63 nM), ROS1, DDR1/2 (IC50=0.1/7 nM) and MKNK1/2 (IC50=7 nM).

Specifications

Chemical Name or Material Merestinib dihydrochloride
CAS 1206801-37-7
Color Pale Yellow
Purity Grade Notes Research
Molecular Formula C30H24Cl2F2N6O3
Quantity 100 mg
Synonym LY2801653 dihydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (159.88 mM)
SMILES [H]Cl.[H]Cl.O=C(C1=CC=C(C)N(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=CC5=C(N(C)N=C5)C=C4C6=CNN=C6)C(F)=C3
Molecular Weight (g/mol) 625.45
Formula Weight 625.45
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.