missing translation for 'onlineSavingsMsg'
Learn More

Mercury Nitric Acid solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12990454 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12990454 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12990454 Supplier Fisher Chemical Supplier No. RP 121 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Glass Bottle
Analyte or Component Names Mercury, nitric acid solution of.
Purity Grade Notes EU Ph Ref: 1052801
Element Me
Quantity 100 mL
Grade Eu. Ph.
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury,nitric acid solution of. European Pharmacopoeia Ref: 1052801 (RP 121)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P391-Collect spillage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.