missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mercury Monoclonal Antibody (1H7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549688
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Mercury is a naturally occurring element that can be found in air, water, and soil. It is a toxic substance that can cause serious health problems, including damage to the nervous system, kidneys, and lungs. Mercury can enter the environment through natural processes, such as volcanic activity and erosion, as well as through human activities, such as mining and industrial processes. It can accumulate in the food chain, particularly in fish and seafood, which can pose a risk to human health.
Specifications
| Mercury | |
| Monoclonal | |
| 3.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 1H7 | |
| Unconjugated | |
| Hg | |
| Conjugate of Hg hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction