missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) trifluoromethanesulfonate, 98%

Product Code. 11313718
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313718 1 g 1g
11393708 5 g 5g
11303718 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313718 Supplier Thermo Scientific Alfa Aesar Supplier No. 039305.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49540-00-3 | C2F6HgO6S2 | 498.718 g/mol

Hg(OTf)2 exhibits remarkable catalytic activity for the hydroxylative cyclization of 1,6-enynes. Mercuric triflate is a very versatile reagent and has been used for several organic catalytic transformations including C-C bond forming cyclizations, alkyne hydrations, heterocycle synthesis and very recently in C-N bond forming reactions. Commonly used as the metal source for Lewis acid catalyzed asymmetric reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49540-00-3
Molecular Formula C2F6HgO6S2
Molecular Weight (g/mol) 498.718
MDL Number MFCD00144746
InChI Key BPVYMDMPLCOQPJ-UHFFFAOYSA-L
Synonym mercury ii trifluoromethanesulfonate, mercury triflate, mercury 2+ ditriflate, hg otf 2, ksc491a8p, mercury ii trifluoromethanesulphonate, mercury 2+ ; trifluoromethanesulfonate, mercury 2+ ion ditrifluoromethanesulfonate, mercury ii trifluoromethanesulfonate trace metals basis
PubChem CID 2775250
IUPAC Name mercury(2+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Hg+2]

Specifications

Melting Point >350°C
Color White
Linear Formula Hg(CF3SO3)2
Quantity 1 g
UN Number UN2025
Sensitivity Moisture sensitive
Solubility Information It is soluble in H2O and CH3CN,fairly soluble in CH3NO2 and CH2Cl2,insoluble in hexane,ether,and toluene.
Formula Weight 498.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercury(II) trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.