missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) trifluoroacetate, 98+%

Product Code. 11379199
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379199 25 g 25g
11369199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379199 Supplier Thermo Scientific Alfa Aesar Supplier No. 088132.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13257-51-7 | C4F6HgO4 | 426.622 g/mol

Mercury(II) trifluoroacetate promotes selective cyclization of methylthio-substituted divinyl ketones to 5-fluorocyclopentenone derivatives. It is a raw material of fine chemicals and pharmaceutical intermediates. So it can be used in the pharmaceutical synthesis. Both linear and cross-linked copolymers of N-vinylcarbazole and methylacrylate can be prepared and mercuriated by using mercury trifluoroacetate in THF or dichloromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13257-51-7
Molecular Formula C4F6HgO4
Molecular Weight (g/mol) 426.622
MDL Number MFCD00013201
InChI Key WMHOESUUCMEQMS-UHFFFAOYSA-L
Synonym mercury ii trifluoroacetate, mercury bis trifluoroacetate, mercury 2+ ditrifluoroacetate, mercury 2+ ; 2,2,2-trifluoroacetate, bis trifluoroacetato mercury, hg ococf3 2, hg o2ccf3 2, mercury bis-trifluoroacetate, mercury ii bis trifluoroacetate
PubChem CID 2723924
IUPAC Name mercury(2+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Hg+2]

Specifications

Melting Point 167°C to 169°C
Color White
Linear Formula Hg(OOCCF3)2
Quantity 25 g
UN Number UN2025
Beilstein 3633860
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 426.62
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Mercury (II) trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.