missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Thiocyanate, Extra Pure, SLR, Fisher Chemical

Product Code. 10607422 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607422 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10607422 Supplier Fisher Chemical Supplier No. M/3300/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 165°C
CAS 592-85-8
Color White
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C4HgK2N4S4
MDL Number 11039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
Molecular Weight (g/mol) 511.10
Formula Weight 316.75g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P330-Rinse mouth.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.