missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) thiocyanate

Product Code. 11384277
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384277 250 g 250g
11394277 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 11384277 Supplier Thermo Scientific Alfa Aesar Supplier No. 037106.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 592-85-8 | C4HgK2N4S4 | 511.10 g/mol

Mercury(II) thiocyanate is used as a precursor to potassium tris(thiocyanato)mercurate(II) and cesium tris(thiocyanato)mercurate(II). It is also used in the determination of chloride ions in water by UV-visible spectroscopy. Further, it acts as a catalyst for the addition of thiocyanic acid to alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
MDL Number MFCD00011039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Melting Point ∽165°C (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 250 g
Beilstein 3561344
Solubility Information Soluble in dilute hydrochloric acid,potassium cyanide and ammonia. Slightly soluble in alcohol and ether. Insoluble in water.
Formula Weight 316.75
Odor Odorless
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 3.71
Chemical Name or Material Mercury(II) thiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.