missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) telluride, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11379219
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379219 25 g 25g
11369219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379219 Supplier Thermo Scientific Alfa Aesar Supplier No. 088244.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mercury telluride in the photoelectromagnetic-Nernst configuration exhibit spectral and frequency responses indicative of thermal rather than photon behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-90-5
Molecular Formula HgTe
Molecular Weight (g/mol) 328.19
MDL Number MFCD00016133
InChI Key VCEXCCILEWFFBG-UHFFFAOYSA-N
Synonym mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
PubChem CID 82914
IUPAC Name tellanylidenemercury
SMILES [Te]=[Hg]

Specifications

Melting Point 673°C
Color Black to Gray
Physical Form Powder
Quantity 25 g
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 328.19
Percent Purity 99%
Odor Odorless
Density 8.63 g/mL
Assay (metals basis)
Chemical Name or Material Mercury (II) telluride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.