missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfide, Thermo Scientific Chemicals

Product Code. p-4412848
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11366858

Brand: Thermo Scientific Alfa Aesar 013483.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mercury(II) sulfide, black is used chiefly as a pigment in paints, rubber and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
SMILES [S--].[Hg++]

Specifications

Melting Point 583.5° (sublimation)
Color Black
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name mercury(2+) sulfanediide
Formula Weight 232.65
Odor Odorless
Appearance Black powder
Density 7.70 g/cm3
Chemical Name or Material Mercury(II) sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.